Anti-EIF2AK3 Rabbit Recombinant Antibody [clone: R09-1F9]
Katalognr. ABNORAB01234.100UL
Supplier: Abnova
Specifications
- Antigen symbol:EIF2AK3
- Clonality:Recombinant
- Clone:R09-1F9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human EIF2AK3.
- Pk:100 µl