Anti-DR5 Mouse Recombinant Antibody [clone: TRA-8]
Catalog # ABNORAB01153.200UG
Supplier: Abnova
Specifications
- Antigen symbol:DR5
- Clonality:Recombinant
- Clone:TRA-8
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human DR5.
- Pk:200 µG