Anti-IL-2R Mouse Recombinant Antibody [clone: DACLIZUMAB]
Catalog # ABNORAB01169.200UG
Supplier: Abnova
Specifications
- Antigen symbol:IL-2R
- Clonality:Recombinant
- Clone:DACLIZUMAB
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human IL-2R.
- Pk:200 µG