Anti-CD11a Mouse Recombinant Antibody [clone: HU1124 (EFALIZUMAB)]
Catalog # ABNORAB01131.200UG
Supplier: Abnova
Specifications
- Antigen symbol:CD11a
- Clonality:Recombinant
- Clone:HU1124 (EFALIZUMAB)
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human CD11a.
- Pk:200 µG