Technische Daten
- Antigen-Symbol:PLK1
- Klonalität:Monoklonal
- Klon:C2
- Konjugation:PE (Phycoerythrin)
- Wirt:Rabbit
- Isotyp:
- Reaktivität:Human
- Environmentally Preferable:
- Modification:pThr210
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human PLK1.
- VE:100 AU