Order Entry
Switzerland
ContactUsLinkComponent
Silicon Cutter, ARGENTAX RAPID
Silicon Cutter, ARGENTAX RAPID
Catalog # MARO594.03
Supplier:  MARTOR ARGENTAX
Silicon Cutter, ARGENTAX RAPID
Catalog # MARO594.03
Supplier:  MARTOR ARGENTAX

Specifications

  • Description:
    Argentax rapid cutter for silicone, 131L×15,5W×43,5H mm
  • Material:
    Plastic
  • Pk:
    1

Specifications

About this item

The ARGENTAX RAPID has found its purpose: silicon cartridges. It has two blades to deal with them. One small pointed one, that you can use to scratch and cut open the plug. And a rectangular one which caps the spout opening like a guillotine according to the thickness you require.

  • Ergonomic
  • For left and right handers
  • Has lanyard hole
  • Practical eyelet
  • The sharp edges of the second blade are best stored in the handle

It has protective cap to ensure that personal transporting is safe, it covers the pointed blades.

Weight: 42,5 g