ContactUsLinkComponent
Experimentation kits, in search of the 'Kissing Disease'
Experimentation kits, in search of the 'Kissing Disease'
Catalog # EDVO274
Supplier:  EDVOTEK
undefined
Experimentation kits, in search of the 'Kissing Disease'
Catalog # EDVO274
Supplier:  EDVOTEK
Specifications
  • Description:
    In Search of the "Kissing Disease"
  • Pk:
    1
Specifications
About this item
Infectious mononucleosis is commonly known as the 'kissing disease'. The causative agent is Epstein-Barr virus (EBV), which can be transmitted through saliva during kissing. In this experiment, students search for the presence of EBV using the ELISA reaction to detect specific viral proteins.
  • For 10 lab groups
  • Requires 50 minutes
Also required: 37 °C incubator, automatic micropipettes with tips.
Delivery information: Kit includes instructions, samples, antigens and antibodies, various solutions and reagents, pipettes and microtest tubes.