ContactUsLinkComponent
Genes in a Tube™
Genes in a Tube™
Catalog # EDVO119
Supplier:  EDVOTEK
undefined
Genes in a Tube™
Catalog # EDVO119
Supplier:  EDVOTEK
Specifications
  • Description:
    Genes in a Tube™
  • Pk:
    1
Specifications
About this item
Kit for teaching students how to extract and precipitate their own DNA. Students can transfer their DNA to a tube that can be used as a pendant on a necklace.
  • For 25 students, plus 1
  • Complete in 30 minutes
Also required: Ice cold alcohol (95% ethanol or isopropyl alcohol) and a 56 °C water bath.
Also required: Ice cold alcohol (95% ethanol or isopropyl alcohol) and a 56 °C water bath.
Delivery: Kit includes instructions, lysis buffer, NaCl solution, Protease, Tris buffer, Methylene Blue Plus solution, microcentrifuge tubes, sterile cotton tipped applicators, transfer pipettes, tubes for DNA precipitation, Gene Tubes, and string.