96- and 384-well cell culture microplates, Advanced TC™
Supplier: Greiner Bio-One
Certificates
About this item
Microplates with a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA.
- Increased cell adherence
- Morphology similar to in vivo conditions
- Optimal cultivation conditions for sensitive cells
- Enables reduced quantities of serum or serum-free use
- Free of cytotoxic substances
Specifications
- Catalog No:
- 392-0227
- 392-0265
- 392-0229
- 392-0269
- 392-0264
- 392-0230
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)