Order Entry
Switzerland
ContactUsLinkComponent
Brady® SPC® Absorbents QUICKWIK Universal Granular Absorbent
Brady® SPC® Absorbents QUICKWIK Universal Granular Absorbent
Catalog # BRDY152275
Supplier:  Brady
undefined
Brady® SPC® Absorbents QUICKWIK Universal Granular Absorbent
Catalog # BRDY152275
Supplier:  Brady

Specifications

  • Type:
    Universal sorbent
  • Description:
    QUICKWIK universal granular absorbent
  • W×D×H:
    33×125×58 mm
  • Absorbency:
    1248 L
  • Pk:
    48

Specifications

About this item

Take care of industrial spills, leaks and drips with QUICKWIK™ universal granular absorbent – more than 4× the absorption of clay granular and produces no carcinogens or dust.

  • Premium absorbent made of 100% recycled material
  • Effectively absorbs hydrocarbons, water-based fluids and most aggressive chemicals
  • Certified safe for landfill disposal
  • Lightweight and easy to deploy
Packaging: Supplied in 50 litres bags, 48 bags per pallet.