TeleScoop Water Samplers
Supplier: Sampling Systems
Certificates
About this item
TeleScoop water sampler is the ultimate extendable water sampler for liquid and water sampling.
- Range of telescopic poles upto 6 m in length
- Robust and lightweight design
- Multi angle elbow design
- Quick and easily interchangeable accessories - no tools needed
- Variety of compatible sampling accessories
- Ideal for sampling from river banks or overhead gantries
The TeleScoop water sampler is the latest extendable sampler for sampling waters and liquids from hard-to-reach places. The extendable rods reach upto 6 m in length and the robust yet lightweight design means easy handling even for the longest of rods. With a dual locking system and multi angle adaptor elbow, the numerous attachments can be preset to angles for effective sampling. Furthermore, the attachments are quick and easily changed without the need for using additional tools. The TeleScoop water sampler by Sampling Systems is the ultimate in long range sampling.
Frequently Bought Together