Kits: restriction enzyme cleavage patterns of DNA, Ready-to-Load™
Supplier: EDVOTEK
Certificates
About this item
Plasmid and lambda DNA are pre-digested with restriction endonucleases that recognise and cut double-stranded DNA within or near defined base sequences. Digests are separated by agarose gel electrophoresis.
- For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Delivery information: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cyclinder and microtipped transfer pipettes.
Frequently Bought Together