CHO Host Cell Protein Standards
Catalog # ENZOENZPRT1220025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 µG
- Conjugation:Unconjugated
- Source:CHO cells
- Seq:02
- Storage conditions:–20 °C
- Protein/peptide name:CHO host cell protein standards
- Formulation:Liquid. In PBS only
- Shipping temperature:20
Specifications
About this item
Quantitative protein standards for Western blotting internal controls or ELISA standards.
CHO host cell protein standards are native proteins that can be used as an internal control for the Western blot analysis of CHO Host Cell Proteins or as standards in an ELISA.