ContactUsLinkComponent
LDPE Bucket, 610/615
LDPE Bucket, 610/615
Codice catalogo KAUT2000070588
Nome del fornitore:  Kautex
undefined
LDPE Bucket, 610/615
Codice catalogo KAUT2000070588
Nome del fornitore:  Kautex
Specifiche
  • Capacity:
    10 L
  • Description:
    LDPE bucket with spout
  • Dimensions:
    193×280×300 mm
  • Pk:
    1
Specifiche
Informazioni su questo articolo
All buckets have strong walls and are very stable for a long service life.
  • Thick-walled, stable industrial and laboratory bucket
  • Stainless steel carrying handle covered with plastic
  • Interior graduations
  • Halal certified
Buckets feature a a liter scale on the inside. The laboratory buckets with spout have a steel carrying handle covered with plastic, and are suitable for food contact.