ContactUsLinkComponent
Pipette jar
  BELAF171300018
Pipette jar
  BELAF171300018
  • Description:
    Pipette jar, white PE, Ø 152 × 460 mm (6 × 18")
  • Pk:
    1
Store and soak pipettes in these leakproof, molded polyethylene pipette jars.
Withstands all common cleaning solutions at room temperature, including sulfuric acid/dichromate.