CRS ZPure™ O₂/H₂O Gas Filters
Supplier: Trajan Scientific and Medical
Certificates
About this item
ZPure™ O₂/H₂O gas filters removes oxygen and water from inert gases, He, Ar, N₂ and H₂, making it ideal for use with GC and GC/MS carrier gas lines. It differs from the Zpure dual filter in that it is completely non-hazardous and is recommended for any application requiring ultra-pure gas, free from oxygen and water.
- High efficiency in-line traps with outstanding capacity
- High quality activated adsorbents for long purifier life and efficient contaminant removal
- Various size and fitting configurations to fit existing installations
- Filter lifetime is dependent on quality of incoming gas and the flow rate
- Individually leak-tested
Efficiency: O₂: <5 ppb, H₂O: <20 ppb
Specifications
- Catalog No:
- SGEA202296-B
- SGEA202295XL-SS
- SGEA202296D-B
- SGEA202295L-SS
- SGEA202295XL-B
- SGEA202295-SS
- SGEA202296L-B
- SGEA202295D-SS-QC
- SGEA202ROHXL-SS-QC
- SGEA202296D-SS
- SGEA202295D-B-QC
- SGEA202ROHXL-B-QC
- SGEA202295L-SS-QC
- SGEA202ROH-SS-QC
- SGEA202296L-SS
- SGEA202295L-B-QC
- SGEA202ROHL-SS-QC
- SGEA202ROHL-B-QC
- SGEA202ROHD-SS-QC
- SGEA202295-B-QC
- SGEA202ROHD-B-QC
- SGEA202ROH-B-QC
- SGEA202295-B
- SGEA202296XL-B
- SGEA202295XL-B-QC
- SGEA202296-SS
- SGEA202295-SS-QC
- SGEA202296-B-QC
- SGEA202296XL-SS
- SGEA202295XL-SS-QC
- SGEA202295D-B
- SGEA202296L-SS-QC
- SGEA202296L-B-QC
- SGEA202295L-B
- SGEA202296D-SS-QC
- SGEA202295D-SS
- SGEA202296D-B-QC
- SGEA202296XL-SS-QC
- SGEA202296XL-B-QC
- SGEA202296-SS-QC
- SGEA202RO2-SS-QC
- SGEA202RO2-B-QC
- Max. flow rate:
- 1,4 L/min
- 7,7 L/min
- 5,2 L/min
- 4,9 L/min
- 7,7 L/min
- 1,4 L/min
- 4,9 L/min
- 5,2 L/min
- 7,7 L/min
- 5,2 L/min
- 5,2 L/min
- 7,7 L/min
- 4,9 L/min
- 1,4 L/min
- 4,9 L/min
- 4,9 L/min
- 4,9 L/min
- 4,9 L/min
- 5,2 L/min
- 1,4 L/min
- 5,2 L/min
- 1,4 L/min
- 1,4 L/min
- 7,7 L/min
- 7,7 L/min
- 1,4 L/min
- 1,4 L/min
- 1,4 L/min
- 7,7 L/min
- 7,7 L/min
- 5,2 L/min
- 4,9 L/min
- 4,9 L/min
- 4,9 L/min
- 5,2 L/min
- 5,2 L/min
- 5,2 L/min
- 7,7 L/min
- 7,7 L/min
- 1,4 L/min
- 1,4 L/min
- 1,4 L/min
- Nominal flow rate:
- 430 ml/min
- 2,5 L/min
- 1,6 L/min
- 1,3 L/min
- 2,5 L/min
- 430 ml/min
- 1,3 L/min
- 1,6 L/min
- 2,5 L/min
- 1,6 L/min
- 1,6 L/min
- 2,5 L/min
- 1,3 L/min
- 430 ml/min
- 1,3 L/min
- 1,3 L/min
- 1,3 L/min
- 1,3 L/min
- 1,6 L/min
- 430 ml/min
- 1,6 L/min
- 430 ml/min
- 430 ml/min
- 2,5 L/min
- 2,5 L/min
- 430 ml/min
- 430 ml/min
- 430 ml/min
- 2,5 L/min
- 2,5 L/min
- 1,6 L/min
- 1,3 L/min
- 1,3 L/min
- 1,3 L/min
- 1,6 L/min
- 1,6 L/min
- 1,6 L/min
- 2,5 L/min
- 2,5 L/min
- 430 ml/min
- 430 ml/min
- 430 ml/min
- Ø×L:
- 32×280 mm
- 50×500 mm
- 50×350 mm
- 38×570 mm
- 50×500 mm
- 32×280 mm
- 38×570 mm
- 50×410 mm
- —
- 50×350 mm
- 50×410 mm
- —
- 38×630 mm
- —
- 38×570 mm
- 38×630 mm
- —
- —
- —
- 32×340 mm
- —
- —
- 32×280 mm
- 50×500 mm
- 50×560 mm
- 32×280 mm
- 32×340 mm
- 32×340 mm
- 50×500 mm
- 50×560 mm
- 50×350 mm
- 38×630 mm
- 38×630 mm
- 38×570 mm
- 50×410 mm
- 50×350 mm
- 50×410 mm
- 50×560 mm
- 50×560 mm
- 32×340 mm
- —
- —
- Max. pressure:
- 68,9 bar
- 13,8 bar
- 13,8 bar
- 68,9 bar
- 13,8 bar
- 68,9 bar
- 68,9 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 68,9 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 68,9 bar
- 13,8 bar
- 13,8 bar
- 68,9 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 68,9 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
- 13,8 bar
Specifications
Frequently Bought Together