SPE Column, NBE TRACE-N20
TCAT611-0051R-NBE
: Tecan
- Particle Size:20 µm
- Capacity:1 ml
- Description:Column NBE TRACE-N20
- Environmentally Preferable:
- Bed weight:5 mg
- Phase:TRACE-N20
- Pk:96
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
- Reversed-phase
- Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.