Order Entry
Switzerland
ContactUsLinkComponent
Thermo Scientific™ Aquanex™ Ultrapure Water Systems
Zertifikate
undefined
Thermo Scientific™ Aquanex™ Ultrapure Water Systems

Technische Daten

  • Catalog No:
  • BARN7201031
  • BARN7101010
  • BARN7101011
  • BARN7201030
  • Feed water:
  • Tap water
  • Tap water
  • Tap water
  • Tap water
  • Flow rate:
  • 2 L/min. max.
  • 2 L/min. max.
  • 2 L/min. max.
  • 2 L/min. max.
  • TOC content:
  • <3 ppb
  • <3 ppb
  • <3 ppb
  • <3 ppb
  • Resistivity at +25 °C:
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • Bacteria:
  • <1 CFU/1000 ml
  • <1 CFU/1000 ml
  • <1 CFU/1000 ml
  • <1 CFU/1000 ml
  • Particles:
  • <1/ml at 0,2 µm
  • <1/ml at 0,2 µm
  • <1/ml at 0,2 µm
  • <1/ml at 0,2 µm
  • W×D×H:
  • 610×406×623 mm
  • 610×406×727 mm
  • 610×406×728 mm
  • 610×406×622 mm
  • Weight:
  • 31 kg
  • 32 kg
  • 32 kg
  • 31 kg

Technische Daten

Frequently Bought Together