Cerex® TRACE-B SPE Columns
Supplier: Tecan
Jump to:
About this item
Strong cationic exchange chemistry ideal for aggressive cleanup of basic target compounds.
- Ion-exchange
- Liquid handler compatible
A 15 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.
4 Options Available Below
Product Details & Documents
Strong cationic exchange chemistry ideal for aggressive cleanup of basic target compounds.
- Ion-exchange
- Liquid handler compatible
A 15 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.
Product Family Options
Product Information
- Particle SizeCapacityBed weightAvailabilityPrice
- TCAT711-0101RColumn Cerex TRACE-B
- TCAT711-0201RColumn Cerex TRACE-B
- TCAT711-335CColumn Cerex TRACE-B
- TCAT711-0506CColumn Cerex TRACE-B