Cerex® TRACE-N20 SPE Column
Catalog # TCAT611-0101R
Supplier: Tecan
Specifications
- Particle Size:15 µm
- Capacity:1 ml
- Description:Column Cerex TRACE-N20
- Endcapped:Yes
- Bed weight:10 mg
- Phase:TRACE-N20
- Pk:96
Specifications
About this item
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
- Reversed-phase
- Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.