Orders LinkContactUsLinkComponent
Cerex® THC SPE Columns
Cerex® THC SPE Columns
Supplier:   Tecan
Cerex® THC SPE Columns
Supplier:  Tecan
Certificates
Cerex® THC SPE Columns
4 Options Available Below
Product Details & Documents
Polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Specially formulated for THC/THCA testing.
Product Family Options
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • TCAT682-0101R
    Cerex Atlas® column THC
    Cerex Atlas® column THC
    THC
    80 Å
    17 µm
    1 ml
    10 mg
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST
  • TCAT682-0353C
    Cerex Atlas® column THC
    Cerex Atlas® column THC
    THC
    80 Å
    17 µm
    3 ml
    35 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • TCAT682-0503C
    Cerex Atlas® column THC
    Cerex Atlas® column THC
    THC
    80 Å
    17 µm
    3 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • TCAT682-0506C
    Cerex Atlas® column THC
    Cerex Atlas® column THC
    THC
    80 Å
    17 µm
    6 ml
    65 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST