Cerex® WWP2 SPE Columns
: Tecan
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
- Reversed-phase
- Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.
Frequently Bought Together