ContactUsLinkComponent
Multicolor and Cut, Sign and Label Printers, S3700
Multicolor and Cut, Sign and Label Printers, S3700
Supplier:  Brady
Certificates
undefined
Multicolor and Cut, Sign and Label Printers, S3700
Supplier:  Brady
Specifications
  • Catalog No:
  • BRDY176428
  • BRDY176430
  • BRDY176429
  • BRDY176432
  • BRDY176431
  • BRDY176427
  • Connector type:
  • Standalone use, USB, ethernet
  • Standalone use, USB, ethernet
  • Ethernet, standalone use, USB, Wi-Fi, Bluetooth™, 5 low energy (Class II)
  • Standalone use, USB, ethernet
  • Ethernet, standalone use, USB, Wi-Fi, Bluetooth™, 5 low energy (Class II)
  • Ethernet, standalone use, USB, Wi-Fi, Bluetooth™, 5 low energy (Class II)
  • Display:
  • Color LCD touchscreen
  • Color LCD touchscreen
  • Color LCD touchscreen
  • Color LCD touchscreen
  • Color LCD touchscreen
  • Color LCD touchscreen
  • Electrical:
  • 110/240 V
  • 110/240 V
  • 110/240 V
  • 110/240 V
  • 110/240 V
  • 110/240 V
  • Power supply:
  • AC adaptor
  • AC adaptor
  • AC adaptor
  • AC adaptor
  • AC adaptor
  • AC adaptor
  • Print method:
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Print resolution:
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • Print speed:
  • 127 mm/s
  • 127 mm/s
  • 127 mm/s
  • 127 mm/s
  • 127 mm/s
  • 127 mm/s
  • Weight:
  • 16,74 kg
  • 16,74 kg
  • 16,74 kg
  • 16,74 kg
  • 16,74 kg
  • 16,74 kg