Chemical-Resistant Cryogenic Polypropylene Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Anbieter: Brady
Zertifikate
Über diesen Artikel
Read text and scan barcodes easily with high-performance B-7425 chemical and temperature resistant polypropylene that resists harsh lab processes.
- Specialized – Labels outlast extremes and are ideal for laboratory identification on vials, centrifuge tubes and test tubes
- Durable – Labels resist chemicals, liquid nitrogen, other harsh environments and a high/low temperature range of –80 to +70 °C
- Adhesive – Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces like glass, copper and stainless steel
- Autoclavable
Ideal for component and equipment labelling, general identification, laboratory labelling, and temperature indicating labelling.
Matte finish, chemical-resistant, cold-resistant, high and low temperature-resistant, smear-resistant, solvent-resistant, indoor use B-7425 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).
Frequently Bought Together