ContactUsLinkComponent
SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator
SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator
Supplier:  THERMO SAVANT
Certificates
undefined
SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator
Supplier:  THERMO SAVANT
Specifications
  • Catalog No:
  • THSASRF110P1-230
  • THSASRF110P2-230
  • W×D×H:
  • 560×370×430 mm
  • 560×370×430 mm
  • Weight:
  • 43 kg
  • 43 kg