ContactUsLinkComponent
AlphaTec® 04-004 Food Processing Gloves
AlphaTec® 04-004 Food Processing Gloves
Supplier:  Ansell
Certificates
AlphaTec® 04-004 Food Processing Gloves
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE04-004/10
  • ANSE04-004/9
  • ANSE04-004/8
  • ANSE04-004/11
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Material:
  • PVC/nitrile
  • PVC/nitrile
  • PVC/nitrile
  • PVC/nitrile
  • Finish:
  • Foam finish
  • Foam finish
  • Foam finish
  • Foam finish
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Anti-static:
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +