Order Entry
Switzerland
ContactUsLinkComponent
Charging Devices with and without Cooling Fan for N-Series
Charging Devices with and without Cooling Fan for N-Series
Catalog # NABE631000528
Supplier:  NABERTHERM
Charging Devices with and without Cooling Fan for N-Series
Catalog # NABE631000528
Supplier:  NABERTHERM

Specifications

  • W×D×H ext.:
    330×1100×880/920 mm
  • Description:
    Charging devices with and without cooling fan for models N 31/H, N 41.., N 61.., N 30/..HA, N 60/..HA
  • Pk:
    1

Specifications

About this item

For charging larger workpieces and annealing boxes.

  • 4 casters, freely movable
  • Equipped with a rack at working height for temporary storage
  • Fixing lock for annealing bags (CWK)
  • CWK version with cooling fan (0.2 kW, 230 V)