0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gas Detection Systems
Accessories for Gas Detection System
Accessories for Gas Detection System
Accessories for Gas Detection System
Catalog # DRAE6811535
Supplier:
Dräger
Expand image
Accessories for Gas Detection System
Catalog # DRAE6811535
Supplier:
Dräger
Specifications
Range:
0 - 200 ppm
Description:
XXS OV-A sensor, for use with the X-am 5000/5600 instruments
Pk:
1
Specifications
About this item
Product Details
Product Details