High-voltage gloves, ActivArmr® 96-003
Supplier: Ansell
Certificates
About this item
Goatskin leather protectors, for extended high-voltage glove life and enhanced hand safety.
- Straight cuff with overedge binding fabric
- Robust goatskin leather: Puncture, abrasion and cut resistance
- Protective design: For longer-lasting electrical safety gloves
- Leather composition enhances dexterity, for unencumbered electrical PPE that remains easy to use
- Suitable for wear over Class 1 to 4 electrical insulated gloves
Composed of premium goatskin leather, ActivArmr® 96-003 protectors guard against moderate cut, puncture and abrasion risks, while ensuring additional personal protection for workers engaged in electrical work, either with or near energized components.
Recommended for: Energized electrical, electrical, maintenance.
Certifications: EN 388:2016 (2111B), ISO 9001, ASTM F496, ASTM F696
Packaging: One pair per package.
Caution: Suitable for wear over Class 1 to 4 electrical insulated gloves.
Specifications
- Catalog No:
- ANSE96-003/8
- ANSE96-003/11
- ANSE96-003/12
- ANSE96-003/10
- ANSE96-003/9
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Material:
- Leather, goatskin
- Leather, goatskin
- Leather, goatskin
- Leather, goatskin
- Leather, goatskin
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388:2016 (2111B)
- EN 388:2016 (2111B)
- EN 388:2016 (2111B)
- EN 388:2016 (2111B)
- EN 388:2016 (2111B)