Order Entry
Switzerland
ContactUsLinkComponent
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% (by titrimetric analysis on dry substance) non cGMP-manufactured, bioCERTIFIED™, Bioprocess Grade Micro and Endotoxin Tested
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% (by titrimetric analysis on dry substance) non cGMP-manufactured, bioCERTIFIED™, Bioprocess Grade Micro and Endotoxin Tested
Supplier:  Spectrum Chemical
Certificates
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% (by titrimetric analysis on dry substance) non cGMP-manufactured, bioCERTIFIED™, Bioprocess Grade Micro and Endotoxin Tested
Supplier:  Spectrum Chemical

Specification Test Results

  • ASSAY (TITRATION; DRIED BASIS):
    min.99.5 %
  • APPEARANCE:
    White Powder
  • SOLUBILITY (1M in H2O):
    Clear and complete
  • pKa @ 25°C:
    7.45-7.65
  • pH (5% in water):
    5.0-6.5
  • LOSS ON DRYING, 140°C, 30 MINUTES:
    max. 0.5%
  • RESIDUE ON IGNITION/SULFATED ASH:
    max. 0.1%
  • UV ABSORBANCE @ 260 nm (1M in water):
    max 0,05
  • UV ABSORBANCE @ 280 nm (1M in water):
    max 0,04
  • UV Absorbance @ 290 nm (33% in water):
    max 0,05
  • UV ABSORANCE @ 405 nm (1M in water):
    max 0,01
  • HEAVY METALS:
    max 5 ppm
  • ENDOTOXINS:
    max.0.002 EU/mg
  • BIOBURDEN:
    max.1000 CFU/g
  • BIOBURDEN YEAST AND MOLD COUNT:
    max.100 CFU/g
  • DNase:
    Not detected
  • PROTEASE:
    Not detected
  • RNase:
    Not detected

Specification Test Results

Frequently Bought Together