VisTex SMS Coveralls, Type 5/6
Supplier: Portwest
Certificates
About this item
This SMS coverall features Hi-Vis reflective tape for enhanced visibility at night or in dim lighting conditions. This trilaminate constructed fabric offers a high tensile strength and toughness. It provides a fluid and particulate barrier to protect the worker without hindering mobility and comfort.
- Reflective tape for increased visibility
- Provides an effective barrier against particulates, aerosols and light sprays
- Anti-static
- Designed with a comfort fit
- Elasticated cuffs for a secure fit
- Ergonomically shaped hood for a perfect fit
- External standards label for quick identification of protection levels
- Two-way zip for quick and easy access
- Retail bag which aids presentation for retail sales
- CE-CAT III
Colour: Orange
Weather conditions: Dark - high visibility, reflective clothing
Certifications: EN ISO 13982 Type 5; EN 13034 Type 6; EN 1073-2; EN 1149 -5.
Specifications
- Catalog No:
- PRTWST36ORRM
- PRTWST36ORRL
- PRTWST36ORRXXL
- PRTWST36ORRXL
- PRTWST36ORRXXXL
- Model:
- ST36
- ST36
- ST36
- ST36
- ST36
- Garment application:
- Services
- Services
- Services
- Services
- Services
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Gender:
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- BizTex™ SMS
- BizTex™ SMS
- BizTex™ SMS
- BizTex™ SMS
- BizTex™ SMS