rubis® Tweezers, PTFE-Coated
New Product
Full PTFE-coated (Teflon) tweezers made from stainless steel.
- Made from anti-magnetic and anti-acid stainless steel
- Good heat and chemical resistance
- Excellent for use in Biotech applications
PTFE coating (Teflon) applied on anti-acid/anti-magnetic stainless steel for a smooth and flawless performance. PTFE coating provides a secure grip, and prevents scratches and almost any substance from adhering to the surface of the tweezers.
Frequently Bought Together