Rubis® Tweezers with Plastic Tips
Supplier: OUTILS RUBIS
Certificates
About this item
Stainless steel tweezers with tips made from epoxy resin glass.
- Temperature resistant up to 155 °C
- Excellent for use in biotech applications
- Good electrical insulation
Made from antimagnetic and anti-acid stainless steel.
Frequently Bought Together