Vis-Tex Cut Gloves Long Cuff
Anbieter: Portwest
Zertifikate
Über diesen Artikel
Cut level D glove with PU dipping for high abrasion resistance, the HPPE and Glass Fibre liner also achieves a level 1 for contact heat.
- Level D cut resistance
- 13 gauge liner for a perfect fit
- Smooth PU coating for increased abrasion resistance
- Extended cuff for wrist and forearm protection
- High visibility liner
- Tested for both cut and heat protection
- Palm dipped to increase dexterity and ventilation
- Breathable seamless liner
- ANSI cut level A4
Flexible hi-vis lining for optimum protection and comfort. Extended 10 cm cuff adds additional protection to the wrist and forearm area. Retail tag aids presentation for retail sales.
Colours: Orange/black
Certifications: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4X43D, EN 407: 2020 X1XXXX. ANSI/ISEA 105: 2016 CUT Level A4, ANSI/ISEA 105: 2016 ABRASION Level 6. CE certified.
Technische Daten
- Catalog No:
- PRTWA631O8RXL
- PRTWA631O8RXXL
- PRTWA631O8RL
- PRTWA631O8RXXXL
- PRTWA631O8RM
- PRTWA631O8RS
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Polyester/steel fibre
- Polyester/steel fibre
- Polyester/steel fibre
- Polyester/steel fibre
- Polyester/steel fibre
- Polyester/steel fibre
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Technische Daten
Frequently Bought Together