Order Entry
Switzerland
ContactUsLinkComponent
Control Standard Endotoxin (CSE)
Control Standard Endotoxin (CSE)
Catalog # WAKFCSE4037-5006
CAS Number:  
undefined
Control Standard Endotoxin (CSE)
Catalog # WAKFCSE4037-5006
CAS Number:  

Specifications

  • Pk:
    3.000 Ng
  • Description:
    Control standard endotoxin, 500 ng vial, lyophilised, Endotoxin derived from E. coli UKT-B

Specifications

About this item

The Control Standard Endotoxin (CSE) is a lyophilised product composed of a 500 ng vial of endotoxin, purified from E. coli UKT-B strain that serves as an accurate and dependable standard for endotoxin testing.

  • Endotoxin derived from E. coli UKT-B
  • Can be used to prepare controls and standard curves
  • Potency of CSE supplied to match each lot of LAL
  • Reconstituted CSE can be stored at 2 to 10 °C for 1 month