PYROSTAR™ ES-F MULTI Kit
Anbieter: FUJIFILM WAKO CHEMICALS
Zertifikate
Über diesen Artikel
The PYROSTAR™ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be used either qualitatively as a gel-clot reagent or quantitatively as a Kinetic-Turbidimetric reagent.
- Endotoxin-specific lysate, avoids false positive results from glucans
- Available in multi-tests vials or single-test vials
- Can be used as either a gel-clot or Kinetic-Turbidimetric Assay (KTA) reagent
- (KTA) assays can be performed in tube reader or microplate reader
- Gel-Clot lysate sensitivities range from 0,015 to 0,25 EU/ml
- Available with matched control standard endotoxin (CSE)
- 100 ul sample size when used with tube reader; 50 µl sample size when used with microplate reader
- Measurement system: Toxinometer or MPR
This multi-test vial is designed to be used with the water bath, heat block or the Toxinometer®. With this configuration, the user dispenses 0,1 ml of dissolved LAL reagent into the appropriate reaction tube, then adding in 0,1 ml of the sample. This multi-test vial is ideal for a user testing a larger number of samples.
PYROSTAR™ ES-F reagents are available with a KTA quantitative range of either 0,001 EU/ml to 10 EU/ml, The KTA quantitative range is related to the Gel-Clot sensitivity.
Frequently Bought Together