Order Entry
Switzerland
ContactUsLinkComponent
XCORE UHPLC Columns, Silica
XCORE UHPLC Columns, Silica
Supplier:  ISERA
Certificates
XCORE UHPLC Columns, Silica
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA411-42CK00
  • ISERA411-42CG00
  • ISERA411-42CH00
  • ISERA411-42CD00
  • ISERA411-42CF00
  • ISERA411-42CA00
  • ISERA411-42CZ00
  • ISERA411-42CB00
  • ISERA411-42CV00
  • ISERA411-42CX00
  • ISERA411-42CM00
  • ISERA411-42CU00
  • Phase name:
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • XCORE
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • USP code:
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together