Order Entry
Switzerland
ContactUsLinkComponent
XELA UHPLC Columns, C4+
XELA UHPLC Columns, C4+
Supplier:  ISERA
Certificates
XELA UHPLC Columns, C4+
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA211-E31M00
  • ISERA211-E31U00
  • ISERA212-E31A30
  • ISERA211-E31K00
  • ISERA211-E31G00
  • ISERA211-E31H00
  • ISERA211-E31D00
  • ISERA211-E31F00
  • ISERA211-E31A00
  • ISERA211-E31Z00
  • ISERA211-E31B00
  • ISERA211-E31V00
  • ISERA211-E31X00
  • Description:
  • Analytical columns
  • Analytical columns
  • Guard column
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Phase name:
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • Endcapped:
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • USP code:
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • L26
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Guard column holders

Product Information

Frequently Bought Together