ContactUsLinkComponent
Masterflex® I/P® Rapid-Load® Pump Heads for Precision and High-Performance Precision Tubing
undefined
Masterflex® I/P® Rapid-Load® Pump Heads for Precision and High-Performance Precision Tubing
  • Model:
  • 77603-20
  • 77603-22
  • Gear ratio:
  • 1:1
  • 1:1
  • No. of channels:
  • 1
  • 1
  • No. of rollers:
  • 3
  • 3
  • Occlusion:
  • Fixed
  • Fixed
  • Open-head sensor:
  • Yes
  • Yes
  • Optimum RPM range:
  • 0,1 - 650 min⁻¹
  • 0,1 - 650 min⁻¹
  • Pump series:
  • I/P
  • I/P
  • Suction lift:
  • 8.8 m (29')
  • 8.8 m (29')
  • Tubing retention:
  • Manual
  • Manual
  • Tubing sizes accepted:
  • I/P® 26, I/P® 73, I/P® 82
  • I/P® 70, I/P® 88, I/P® 89
  • Volume per revolution with I/P 26 tubing:
  • 6,7 ml
  • Volume per revolution with I/P 70 tubing:
  • 13,4 ml
  • Volume per revolution with I/P 73 tubing:
  • 13,7 ml
  • Volume per revolution with I/P 82 tubing:
  • 20,8 ml
  • Volume per revolution with I/P 88 tubing:
  • 23,2 ml
  • Volume per revolution with I/P 89 tubing:
  • 32,1 ml
  • Warranty:
  • 2 years
  • 2 years
Frequently Bought Together