Orders LinkContactUsLinkComponent
VWR® Sample Vials
VWR® Sample Vials
VWR® Sample Vials
VWR® Sample Vials
Screw neck vials with flat bottom made from clear borosilicate glass.
  • Highly resistant against chemicals
  • High temperature resistance
  • Delivered with screw caps
Ideal for storage of liquid and powders.
Product Family Options
  • Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Cap material
    Cap colour
    Septum
    Sterile
  • 2,00 ml
    12×35 mm
    Clear
    Yes
    8-425
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 29.70
    SFr. 29.70
    1 * 144 ST
       SFr. 29.70
  • 4,00 ml
    15×45 mm
    Clear
    Yes
    13-425
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 34.00
    SFr. 34.00
    1 * 144 ST
       SFr. 34.00
  • 6,00 ml
    16×50 mm
    Clear
    Yes
    13-425
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 34.00
    SFr. 34.00
    1 * 144 ST
       SFr. 34.00
  • 8,00 ml
    17×60 mm
    Clear
    Yes
    15-425
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 42.30
    SFr. 42.30
    1 * 144 ST
       SFr. 42.30
  • 12,00 ml
    19×65 mm
    Clear
    Yes
    15-425
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 42.30
    SFr. 42.30
    1 * 144 ST
       SFr. 42.30
  • 16,00 ml
    21×70 mm
    Clear
    Yes
    18-400
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 50.90
    SFr. 50.90
    1 * 144 ST
       SFr. 50.90
  • 24,00 ml
    23×85 mm
    Clear
    Yes
    20-400
    PP
    Black
    PTFE liner
    -
    1 * 144 ST
    1 * 144 ST - SFr. 59.20
    SFr. 59.20
    1 * 144 ST
       SFr. 59.20