Permanent vinyl (B-439) is a thermal transfer printable material ideal for applications needing various colours like finished product ID, rating plates, and general-purpose ID.
- Available in many different colours as a custom option
- Designed for use in ambient conditions with limited solvent exposure
- Indoor/outdoor vinyl film has an acrylic pressure sensitive adhesive
- Service temperature of (−94 to 104 °F) −70 to 40 °C
- Gloss finish
Ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 or M61-R4410-WT for the M610/BMP61/Bradyprinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.