Engraved Replcement Labels for M710, M610, M611
Nome del fornitore: Brady
Certificati
Informazioni su questo articolo
Engraved plate replacement labels for the M610 and M611 thermal transfer portable printers. These parts can also be printed on the M710 label printer provided the roll adapter (M71-ADAPT) is used.
- Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
- Polyethylene foam laminate Polyester material
- Service temperature of −40 to +100 °C
- Gloss finish
Ideal for data and telecommunications labelling, electrical labelling, and general identification applications.
Frequently Bought Together