BradyGrip® Print-on-Hook Labels with VELCRO® Hook for M710 and BMP71
Supplier: Brady
Certificates
About this item
For wire and cable ID, use BradyGrip® print-on hook material, an exclusive print-and-place material that can be easily applied, repositioned, or removed.
- Features a printable front and grippy back with tiny hooks
- Attaches to VELCRO® brand ONE-WRAP® strap for easy wire and cable ID
- Will not damage wires and cables like nylon cable ties
- Maximise uptime with faster, printable ID on cable bundles
- Matte finish
Removable and repositionable are ideally suited for wire and cable labelling. A ribbon is required to print this material. Use part numbers M7-R6000 and M71-R6000.
VELCRO® brand ONE-WRAP® product not included with purchase. VELCRO® and ONE-WRAP® are registered trademarks of Velcro IP holdings LLC. Used with permission.
Packaging: One tape per box.
Frequently Bought Together