ContactUsLinkComponent
Gas Chromatograph with FID (GC-FID)
Catalog # CCDL928270-240
Supplier:  LUCIDITY
undefined
Gas Chromatograph with FID (GC-FID)
Catalog # CCDL928270-240
Supplier:  LUCIDITY
Specifications
  • Voltage:
    240 V
  • Description:
    Gas chromatograph with FID
  • W×D×H:
    266,7×457,2×317,5 mm (10,5×18×11,5")
  • Pk:
    1
Specifications
About this item
Compact, economical, fully functional gas chromatograph.
  • Lucidity quick column change for quick and easy column installation
  • Equipped with a split/splitless injection port
  • Contains a very small oven which allows for rapid heating and cooling
  • Intuitive Lucidity Software included with purchase of instrument
  • Instrument changes color to indicate status
  • Self-installable and ease of use design
  • Low power consumption
The Lucidity GC has one of the smallest footprints of any fully functional GC-FID. Changing GC columns in the Lucidity GC is as simple as loading a gaming cartridge. Fused silica and metal capillary columns can be installed into the column holder.
Delivery: The instrument comes with everything required for analysis, including: laptop containg Lucidity Software, starter column, full accessory kit and training videos. Self-installable in under 30 minutes. With a 24-month warranty, this GC-FID will provide reliable results for years to come.
Additional instruments
Product Information