ContactUsLinkComponent
Scotch-Brite™ 350 Extra-Thick Delicate Surface Cleaning Pads
Scotch-Brite™ 350 Extra-Thick Delicate Surface Cleaning Pads
  MMMAHP350
 :  3M
undefined
Scotch-Brite™ 350 Extra-Thick Delicate Surface Cleaning Pads
  MMMAHP350
 :  3M
  • Colour:
    White
  • Description:
    Scotch-Brite™ Extra-Thick delicate surface cleaning pad 350
  • L×W:
    158×98 mm
  • Pk:
    40
Scotch-Brite™ Extra-thick delicate surface cleaning pads 350 are made from a high-quality synthetic fibre with a non-woven open structure. Non-abrasive cleaning particles are spread uniformly throughout the thickness of the fibre and linked with a durable, long-lasting resin.
  • Delicate and non-aggressive on surfaces
  • Flexible and adaptable
  • Long-lasting
  • Easy to rinse
Clean delicate surfaces without leaving scratches. These extra-thick cleaning pads are flexible and adaptable for delicate and non-aggressive cleaning on surfaces.