ContactUsLinkComponent
Scotch-Brite™ 9000 All-Purpose Scouring Pad
Scotch-Brite™ 9000 All-Purpose Scouring Pad
Catalog # MMMASB9000
Supplier:  3M
undefined
Scotch-Brite™ 9000 All-Purpose Scouring Pad
Catalog # MMMASB9000
Supplier:  3M
Specifications
  • Description:
    Scotch-Brite™ All-purpose scouring pad
  • L×W:
    101,6×133,3 mm
  • Pk:
    40
Specifications
About this item
The Scotch-Brite™ All-purpose scouring pad 9000 is a thick, hand-sized scouring pad designed for cleaning most types of surfaces without scratching.
  • Versatile, hand-sized pad for dish scrubbing
  • Can be used with Scotch-Brite™ Multi-Purpose Pad Holder 405-R and 3M™ Twist Lok™ Pad Holder 961
Clean most types of surfaces without scratching with Scotch-Brite™ All-purpose scouring pad 9000. Pads are safe for use on porcelain, ceramic, stainless steel, glass, and non-stick pans. It’s specially designed for cleaning hot griddles with mirror or highly polished finishes. Certified by HACCP International as 'Food Safe' and 'Fit for Purpose'.