Hard Hat Replacement Suspension Systems, G/H
MMMAG2N
: 3M
- Description:Ratchet suspension, plastic sweatband, G2N
- Pk:1
3M™ Ratchet suspension, G2N is a replacement ratchet suspension cradle and plastic sweatband for all 3M™ Hard Hats, except the X5000 series.
- Replacement ratchet suspension cradle with plastic sweatband
- Ratchet adjustment system provides simple and secure size adjustment
- Suitable for use with all 3M™ hard hats
Replace the ratchet suspension and plastic sweatband on a hard hat with 3M™ Ratchet Suspension, G2N. This replacement ratchet suspension cradle is designed for all 3M™ Hard Hats, except the X5000 series. It snaps easily into place to provide all the comfort and easy adjustability of the original system. The adjustment system provides simple and secure size adjustment for head sizes from 54 to 63 cm.