VWR® Square Polycarbonate Media Bottles
Anbieter: VWR Collection
Zertifikate
Über diesen Artikel
VWR® square polycarbonate bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −130 to +121 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.
- Made from durable 100% virgin polycarbonate material
- Suitable for frozen storage down to −130 °C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and closures (HDPE/PP) provided for applicable products
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
Bottles are arranged in shrink-wrapped trays and polybag to prevent contamination. 2 L bottles feature a moulded hand grip for better ergonomics. Bottles 30 ml and above have volume graduations.
Certifications: US FDA 21CRF, USP CLASS VI compliant Resin.
Accessories information: Spare closures are available for specific application requirements. See part numbers VWRI210-000111, VWRI210-000112, VWRI210-000113, VWRI210-000114, VWRI210-000115, VWRI210-000116, VWRI210-000117, VWRI210-000118, VWRI210-000119 and VWRI210-000120.
Packaging: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.
Zugehörige Informationen
Zertifikate
Suche nach ChargennummerCoC VWR PC Bottles Extractables and Leachable CoC VWR PC Bottles Regulatory Compliances CoC VWR® Square Polycarbonate Media Bottles, Avantor COC_VWRI210-000094_V-54-15J-080923 COC_VWRI210-000095_V-54-15J-240823
More
TDS Square PC Bottles
Datasheet
VWR PETG Square Media Bottles VWR® PETG Square Media Bottles
Product Literature
Spare Closures
Produktinformationen
Frequently Bought Together