ContactUsLinkComponent
Cole-Parmer® HM-200 Large Volume Heating Mantles
Cole-Parmer® HM-200 Large Volume Heating Mantles
Supplier:  Cole-Parmer
Certificates
undefined
Cole-Parmer® HM-200 Large Volume Heating Mantles
Supplier:  Cole-Parmer
Specifications
  • Catalog No:
  • ELET36013-22
  • ELET36013-20
  • ELET36013-16
  • ELET36013-18
  • Electrical:
  • 230 V, 50 Hz
  • 230 V, 50 Hz
  • 230 V, 50 Hz
  • 230 V, 50 Hz
  • Max. temperature:
  • 450 °C
  • 450 °C
  • 450 °C
  • 450 °C