WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Clear Glass
Supplier: DWK Life Sciences
Certificates
About this item
Complete range of clear crimp top vials, rubber stoppers, and seals that are sterilised and ready-to-use.
- Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
- Complete manufacturing and certificate of analysis traceability
- Container Closure Integrity (CCI) validation
Ready-to-use, just-in-time, minimum 180 day shelf life from time of shipment.
Certifications: USP 788 / EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP 85 /EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (Vials, Stoppers, and Seals), USP 660 / EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/ EP 3.2.9 Elastomeric Closures used in Parenteral Preparations.
Frequently Bought Together